Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145844.1 | NP_665843.1 | 1 | 163 | ||
GenBank mRNA | BC150715.1 | - | 1 | 262 | 9087626 |
Gene Symbol: Vmn1r200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134212.1 | NP_598973.1 | 1 | 163 | ||
GenBank mRNA | BC131902.1 | - | 1 | 245 | 40130179 | |
BC131904.1 | - | 1 | 245 | 40130181 |
Gene Symbol: Vmn1r206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134216.1 | NP_598977.1 | 1 | 163 | ||
GenBank mRNA | BC132625.1 | - | 1 | 263 | 40130902 | |
BC138067.1 | - | 1 | 263 | 8861086 |
Gene Symbol: Vmn1r207-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166709.1 | NP_001160181.1 | 1 | 163 |
Gene Symbol: Vmn1r209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 163 |
Gene Symbol: Vmn1r214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134214.1 | NP_598975.1 | 1 | 292 | ||
GenBank mRNA | BC145835.1 | - | 1 | 336 | 40131138 |