Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145857.2 | NP_665856.2 | 4, 5 | 2458 | ||
GenBank mRNA | AF520774.1 | - | 4, 5 | 2503 | ||
AK089843.1 | - | 4, 5 | 2422 | |||
AY160220.1 | - | 3, 4 | 2387 | |||
AY160221.1 | - | 4, 5 | 2459 | |||
BC044774.2 | - | 4, 5 | 2485 |