Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270423.1 | NP_001257352.1 | 2 | 401 | ||
GenBank mRNA | CB173711.1 | - | 2 | 408 | ||
CB174210.1 | - | 2 | 401 | |||
CB174322.1 | - | 1 | 235 |
Gene Symbol: Olfr1274-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146263.2 | NP_666375.2 | 1 | 270 | ||
GenBank mRNA | BC145937.1 | - | 270 | 40131240 |
Gene Symbol: Olfr1506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146265.2 | NP_666377.2 | 2 | 401 | ||
GenBank mRNA | BC111438.1 | - | 2 | 121 |