Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005230.2 | NP_001005230.2 | 1 | 573 | ||
GenBank mRNA | BC119447.1 | - | 1 | 573 | 40044257 |