Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011789.1 | NP_001011789.1 | 1 | 532 | ||
GenBank mRNA | BC147220.1 | - | 1 | 607 | 9056253 | |
BC147221.1 | - | 1 | 607 | 9056247 |