Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011790.1 | NP_001011790.1 | 1 | 796 | ||
GenBank mRNA | BC119435.1 | - | 1 | 796 | 40041463 |