Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146838.2 | NP_667049.2 | 2 | 678 |
Gene Symbol: Olfr1126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146837.2 | NP_667048.2 | 1 | 498 |
Gene Symbol: Olfr1129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011836.2 | NP_001011836.2 | 1 | 498 |