Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011855.1 | NP_001011855.1 | 1 | 272 |
Gene Symbol: Olfr170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146957.1 | NP_667168.1 | 1 | 272 | ||
GenBank mRNA | BC132585.1 | - | 1 | 309 | 40130862 | |
BC132587.1 | - | 1 | 309 | 40130864 |
Gene Symbol: Olfr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146958.2 | NP_667169.2 | 1 | 275 |