Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282480.1 | NP_001269409.1 | 5 | 414 | ||
NM_001282481.1 | NP_001269410.1 | 4 | 288 | |||
NM_001282482.1 | NP_001269411.1 | 4 | 288 | |||
NM_001282483.1 | NP_001269412.1 | 4 | 288 | |||
NM_001282485.1 | NP_001269414.1 | 9 | 1339 | |||
NM_006259.2 | NP_006250.1 | 9 | 1339 | |||
XM_005263126.3 | XP_005263183.1 | 9 | 1774 | |||
XM_017008413.1 | XP_016863902.1 | 9 | 1776 | |||
XM_017008414.1 | XP_016863903.1 | 9 | 1771 | |||
XM_017008415.1 | XP_016863904.1 | 9 | 1181 | |||
XM_017008416.1 | XP_016863905.1 | 9 | 1693 | |||
XM_017008417.1 | XP_016863906.1 | 8 | 1157 | |||
XM_017008418.1 | XP_016863907.1 | 8 | 1157 | |||
GenBank mRNA | AK296979.1 | - | 4 | 275 | ||
AK297673.1 | - | 9 | 1286 | |||
AK316140.1 | - | 5 | 414 | |||
D70899.1 | - | 8 | 1144 | |||
DA462272.1 | - | 4 | 288 | |||
DC348270.1 | - | 4 | 277 | |||
X94612.1 | - | 8 | 1158 |