Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002745.4 | NP_002736.3 | 4 | 790 | ||
NM_138957.3 | NP_620407.1 | 4 | 790 | |||
GenBank mRNA | AK090869.1 | - | 5 | 647 | ||
AK295188.1 | - | 6 | 723 | |||
AK307615.1 | - | 5 | 635 | |||
BC017832.1 | - | 4 | 790 | 4685557 | ||
BC099905.1 | - | 4 | 755 | 5264558 | ||
DQ399292.1 | - | 4 | 550 | |||
EF565108.1 | - | 2 | 236 | |||
Z11694.1 | - | 4 | 734 | |||
Z11695.1 | - | 4 | 648 |