Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146879.2 | NP_667090.1 | 2 | 872 | ||
GenBank mRNA | BC120838.1 | - | 1 | 932 | 40129761 | |
BC172163.1 | - | 1 | 850 | |||
BC172164.1 | - | 1 | 850 | |||
BC172166.1 | - | 1 | 850 |
Gene Symbol: Olfr325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207153.2 | NP_997036.2 | 1 | 855 | ||
GenBank mRNA | BC141015.1 | - | 1 | 879 | 9055627 |
Gene Symbol: Olfr329-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147083.1 | - | 1 | 878 | 9056127 | |
BC147084.1 | - | 1 | 878 | 9056147 |