Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207155.2 | NP_997038.2 | 1 | 640 | ||
GenBank mRNA | BC148220.1 | - | 1 | 644 | 40135828 | |
BC148277.1 | - | 1 | 644 | 40135829 |
Gene Symbol: Olfr130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146487.1 | NP_666698.1 | 1 | 640 | ||
GenBank mRNA | BC128267.1 | - | 1 | 640 | 40043327 |