Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 8, 9 | 786 | ||
NR_110887.1 | 8, 9 | 786 | ||||
XM_011530706.1 | XP_011529008.1 | 6, 7 | 510 | |||
GenBank mRNA | AF001008.1 | - | 8, 9 | 685 | ||
AF001174.1 | - | 8, 9 | 691 | |||
AF031135.1 | - | 8, 9 | 700 | |||
AK291845.1 | - | 8, 9 | 737 | |||
AK308969.1 | - | 8, 9 | 702 | |||
BC027933.1 | - | 8, 9 | 743 | 5199628 | ||
BX361670.2 | - | 8, 9 | 769 | |||
BX444192.2 | - | 8, 9 | 784 | |||
CR456514.1 | - | 8, 9 | 687 | |||
U53442.1 | - | 8, 9 | 728 | |||
U92268.1 | - | 8, 9 | 689 | |||
Y14440.1 | - | 8, 9 | 691 |