Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146291.1 | NP_666403.1 | 1 | 52 |
Gene Symbol: Olfr1477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146696.2 | NP_666907.2 | 2 | 204 | ||
GenBank mRNA | CB173171.1 | - | 2 | 204 |
Gene Symbol: Olfr1487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146636.1 | NP_666847.1 | 1 | 52 |