Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146292.1 | NP_666404.1 | 1 | 820 | ||
GenBank mRNA | BC119382.1 | - | 1 | 908 | 8734131 | |
BC120627.1 | - | 1 | 908 | 40129550 |