Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146294.2 | NP_666406.2 | 1 | 211 | ||
GenBank mRNA | BC119292.1 | - | 1 | 211 | 8734041 | |
BC137817.1 | - | 1 | 211 | 8860836 |
Gene Symbol: Olfr1166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146650.2 | NP_666861.2 | 1 | 155 | ||
GenBank mRNA | BC051250.1 | - | 1 | 155 | 6306097 |
Gene Symbol: Olfr1170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146532.1 | NP_666743.1 | 1 | 145 | ||
GenBank mRNA | BC127994.1 | - | 1 | 149 | 40135743 |