Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000682.6 | NP_000673.2 | 1 | 2322 | ||
GenBank mRNA | BC133021.1 | - | 1 | 2322 | 40147043 | |
BC136537.1 | - | 1 | 2322 | 9020527 |
Set Membership: |
G-protein coupled receptor |