Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146319.2 | NP_666431.2 | 1 | 285 | ||
GenBank mRNA | BC132575.1 | - | 1 | 285 | 40130852 | |
BC145712.1 | - | 1 | 285 | 40131015 |
Gene Symbol: Olfr725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146317.2 | NP_666429.2 | 1 | 358 | ||
GenBank mRNA | BC125411.1 | - | 1 | 358 | 40129926 | |
BC125417.1 | - | 1 | 358 | 40129932 |
Gene Symbol: Olfr726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146316.2 | NP_666428.2 | 1 | 259 |