Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002754.4 | NP_002745.1 | 6 | 582 | ||
NR_072996.1 | 6 | 582 | ||||
GenBank mRNA | AB209586.1 | - | 5 | 1540 | ||
AB451221.1 | - | 7 | 483 | |||
AB451344.1 | - | 7 | 483 | |||
AF004709.1 | - | 6 | 531 | |||
AF092535.1 | - | 6 | 504 | |||
AF100546.1 | - | 6 | 485 | |||
AK222922.1 | - | 6 | 474 | |||
AK308018.1 | - | 6 | 563 | |||
BC000433.2 | - | 6 | 524 | 2819932 | ||
BC001641.2 | - | 6 | 524 | 2819932 | ||
BC004428.1 | - | 6 | 524 | 2819932 | ||
BC085196.1 | - | 6 | 552 | 6107307 | ||
BM921729.1 | - | 6 | 583 | |||
BT007221.1 | - | 6 | 483 | |||
CR536490.1 | - | 6 | 483 | |||
CR541687.1 | - | 6 | 483 | |||
DR006449.1 | - | 6 | 586 | |||
KU178281.1 | - | 6 | 483 | |||
KU178282.1 | - | 6 | 483 | |||
U93232.1 | - | 6 | 490 | |||
Y10488.1 | - | 6 | 505 |