Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146339.3 | NP_666451.1 | 3 | 432 | ||
GenBank mRNA | BC046590.1 | - | 3 | 432 | 6309128 |