Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030662.3 | NP_109587.1 | 2 | 463 | ||
XM_006722799.2 | XP_006722862.1 | 2 | 518 | |||
XM_017026989.1 | XP_016882478.1 | 2 | 518 | |||
XM_017026990.1 | XP_016882479.1 | 2 | 518 | |||
XM_017026991.1 | XP_016882480.1 | 2 | 518 | |||
GenBank mRNA | BC000471.2 | - | 2 | 463 | 2961198 | |
BC018645.2 | - | 2 | 405 | 4025987 | ||
BX538081.1 | - | 3 | 213 | |||
L11285.1 | - | 2 | 293 |
Gene Symbol: LOC407835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002144.1 | 1 | 453 |