Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206804.1 | NP_001193733.1 | 4 | 342 | ||
NM_002757.3 | NP_002748.1 | 4 | 926 | |||
NM_145160.2 | NP_660143.1 | 4 | 926 | |||
XM_011521787.2 | XP_011520089.1 | 4 | 1003 | |||
XM_011521788.2 | XP_011520090.1 | 4 | 1003 | |||
XM_017022414.1 | XP_016877903.1 | 4 | 1003 | |||
GenBank mRNA | AK293459.1 | - | 4 | 342 | ||
AK315435.1 | - | 4 | 473 | |||
BC008838.2 | - | 4 | 583 | 4100679 | ||
BT006780.1 | - | 4 | 273 | |||
CR542229.1 | - | 4 | 273 | |||
U71087.1 | - | 4 | 334 | |||
U71088.2 | - | 4 | 278 |