Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146368.1 | NP_666480.1 | 1 | 216 | ||
GenBank mRNA | BC104114.1 | - | 1 | 214 | ||
BC104115.1 | - | 1 | 214 | |||
BC114393.1 | - | 1 | 215 | |||
BC114424.1 | - | 1 | 215 |