Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146413.2 | NP_666525.2 | 1 | 739 | ||
GenBank mRNA | BC106969.1 | - | 1 | 739 | 40041195 | |
BC106970.1 | - | 1 | 737 |
Gene Symbol: Olfr462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146411.2 | NP_666523.2 | 1 | 739 | ||
GenBank mRNA | BC108939.1 | - | 739 | 40041186 | ||
BC108940.1 | - | 740 |