Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146430.2 | NP_666641.2 | 1 | 701 | ||
GenBank mRNA | BC132583.1 | - | 1 | 701 | 40130860 |
Gene Symbol: Olfr743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177508.1 | NP_001170979.1 | 1 | 616 |
Set Membership: |
![]() |