Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146431.2 | NP_666642.2 | 1 | 889 | ||
GenBank mRNA | BC112967.1 | - | 832 | 40043789 | ||
BC114920.2 | - | 832 | 40040851 | |||
BC119377.1 | - | 1 | 889 | 8734126 | ||
BC148301.2 | - | 832 | 40043795 |