Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146432.2 | NP_666643.1 | 1 | 580 | ||
GenBank mRNA | BC133656.1 | - | 1 | 580 | 40042187 |
Gene Symbol: Olfr1513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012269.2 | NP_001012269.2 | 1 | 580 |