Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146465.2 | NP_666676.1 | 1 | 896 | ||
GenBank mRNA | BC132008.1 | - | 1 | 896 | 40130285 | |
BC132012.1 | - | 1 | 896 | 40130289 |
Gene Symbol: Olfr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146511.2 | NP_666722.2 | 1 | 893 | ||
GenBank mRNA | BC141327.1 | - | 1 | 893 | 9055939 | |
BC141329.1 | - | 1 | 893 | 9055941 | ||
BC148321.1 | - | 1 | 820 | 40054936 |
Gene Symbol: Olfr97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146512.1 | NP_666723.1 | 1 | 823 |