Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002761.2 | NP_002752.1 | 1 | 162 | ||
GenBank mRNA | AK311987.1 | - | 1 | 155 | ||
AY651260.1 | - | 1 | 143 | |||
BC003673.1 | - | 1 | 155 | 3935638 | ||
BT006746.1 | - | 1 | 65 | |||
CD360017.1 | - | 1 | 136 | |||
Y00443.1 | - | 1 | 165 |