Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207230.1 | NP_997113.1 | 1 | 133 | ||
GenBank mRNA | BC104264.1 | - | 132 | |||
BC111096.1 | - | 130 | ||||
BC152316.1 | - | 1 | 133 | 40040380 |
Gene Symbol: Olfr318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146501.2 | NP_666712.2 | 1 | 181 | ||
GenBank mRNA | BC137758.1 | - | 1 | 181 | 8860777 |
Gene Symbol: Olfr319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146500.2 | NP_666711.2 | 1 | 133 | ||
GenBank mRNA | BC104265.1 | - | 1 | 133 | 40040340 |