Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146879.2 | NP_667090.1 | 2 | 905 | ||
GenBank mRNA | BC120838.1 | - | 1 | 965 | 40129761 | |
BC172163.1 | - | 1 | 883 | |||
BC172164.1 | - | 1 | 883 | |||
BC172166.1 | - | 1 | 883 |
Gene Symbol: Olfr328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146502.2 | NP_666713.2 | 1 | 864 | ||
GenBank mRNA | BC110465.2 | - | 1 | 864 | 40040838 | |
BC110466.2 | - | 1 | 864 | 40040839 |