Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146504.1 | NP_666715.1 | 1 | 609 | ||
GenBank mRNA | BC127985.1 | - | 1 | 613 | 40135556 |
Gene Symbol: Olfr960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146279.1 | NP_666391.1 | 1 | 615 | ||
GenBank mRNA | BC132468.1 | - | 1 | 635 | 40130745 | |
BC132470.1 | - | 1 | 635 | 40130747 |
Gene Symbol: Olfr963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011827.1 | NP_001011827.1 | 1 | 609 |