Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146511.2 | NP_666722.2 | 1 | 581 | ||
GenBank mRNA | BC141327.1 | - | 1 | 581 | 9055939 | |
BC141329.1 | - | 1 | 581 | 9055941 | ||
BC148321.1 | - | 1 | 508 | 40054936 |