Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195226.1 | NP_001182155.1 | 2 | 130 | ||
NM_016335.4 | NP_057419.4 | 3 | 545 | |||
GenBank mRNA | AB209472.1 | - | 2 | 196 | ||
BC036534.1 | - | 2 | 354 | |||
BC068260.1 | - | 2 | 347 | |||
BC094736.1 | - | 2 | 175 | |||
BC118597.1 | - | 2 | 130 | 40108132 | ||
BC121809.1 | - | 2 | 130 | 40108133 | ||
CN362765.1 | - | 2 | 296 | |||
KJ534920.1 | - | 2 | 341 | |||
U79754.1 | - | 3 | 545 |
Gene Symbol: LOC102724788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724935.3 | XP_006724998.1 | 2 | 374 | ||
XM_006724936.3 | XP_006724999.1 | 2 | 349 | |||
XM_017030166.1 | XP_016885655.1 | 2 | 471 | |||
XM_017030168.1 | XP_016885657.1 | 2 | 192 |