Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000024.5 | NP_000015.1 | 1 | 937 | ||
GenBank mRNA | AK223025.1 | - | 1 | 900 | ||
AK313151.1 | - | 1 | 919 | |||
AY136741.1 | - | 1 | 698 | |||
BC012481.2 | - | 1 | 907 | 4538187 | ||
BC063486.1 | - | 1 | 885 | 5241774 | ||
BC073856.1 | - | 1 | 888 | 5217922 | ||
KJ534762.1 | - | 1 | 698 | |||
M15169.1 | - | 1 | 2285 | |||
X04827.1 | - | 1 | 875 |
Set Membership: |
G-protein coupled receptor |