Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146531.2 | NP_666742.2 | 1 | 235 | ||
GenBank mRNA | BC104084.2 | - | 1 | 235 | 40042096 |