Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207566.1 | NP_997449.1 | 1 | 151 | ||
GenBank mRNA | BC104085.2 | - | 1 | 151 | 40042097 | |
BC104278.1 | - | 1 | 151 | 40041579 | ||
BC104279.1 | - | 1 | 151 | 40041581 | ||
BC127020.1 | - | 1 | 151 | 40042733 | ||
BC134355.1 | - | 1 | 151 | 40041582 |
Gene Symbol: Olfr1168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146531.2 | NP_666742.2 | 1 | 151 | ||
GenBank mRNA | BC104084.2 | - | 1 | 151 | 40042096 |