Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146536.2 | NP_666747.2 | 1 | 969 | ||
GenBank mRNA | BC117002.1 | - | 1 | 969 | 40126321 | |
BC120742.1 | - | 1 | 969 | 40129665 |
Gene Symbol: Olfr311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146537.2 | NP_666748.2 | 1 | 887 |
Gene Symbol: Olfr312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011819.2 | NP_001011819.1 | 1 | 887 | ||
GenBank mRNA | BC104268.1 | - | 1 | 887 | 40040766 | |
BC104273.1 | - | 1 | 887 | 40041031 |