Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146560.2 | NP_666771.2 | 2 | 200 | ||
GenBank mRNA | BC120630.1 | - | 1 | 118 | 40129553 | |
BC120632.1 | - | 1 | 118 | 40129555 |
Gene Symbol: Olfr869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146557.2 | NP_666768.2 | 2 | 174 |