Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146578.2 | NP_666789.1 | 3 | 385 | ||
GenBank mRNA | AK035445.1 | - | 2 | 267 | ||
AK041245.1 | - | 3 | 385 |