Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146597.2 | NP_666808.1 | 2 | 1115 | ||
GenBank mRNA | BC120573.1 | - | 1 | 895 | 40129496 |