Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146623.1 | NP_666834.1 | 1 | 580 | ||
GenBank mRNA | BC119361.1 | - | 1 | 665 | 8734110 | |
BC119363.1 | - | 1 | 665 | 8734112 |