Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146663.2 | NP_666874.2 | 1 | 746 | ||
GenBank mRNA | BC116806.1 | - | 1 | 746 | 40126125 | |
BC116808.1 | - | 1 | 746 | 40126127 |
Gene Symbol: Olfr732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146665.2 | NP_666876.1 | 1 | 743 | ||
GenBank mRNA | BC120848.1 | - | 1 | 743 | 40129771 |