Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031849.2 | NP_001027019.1 | 5 | 997 | ||
NM_001879.5 | NP_001870.3 | 5 | 997 | |||
NM_139125.3 | NP_624302.1 | 5 | 997 | |||
NR_033519.1 | 4 | 765 | ||||
XM_006713701.2 | XP_006713764.1 | 6 | 1070 | |||
XM_011512989.2 | XP_011511291.1 | 5 | 991 | |||
XM_011512990.2 | XP_011511292.1 | 5 | 991 | |||
XM_011512991.2 | XP_011511293.1 | 5 | 986 | |||
XM_017006869.1 | XP_016862358.1 | 6 | 1075 | |||
XM_017006870.1 | XP_016862359.1 | 4 | 963 | |||
XM_017006871.1 | XP_016862360.1 | 5 | 991 | |||
XM_017006872.1 | XP_016862361.1 | 5 | 850 | |||
GenBank mRNA | AF284421.1 | - | 5 | 697 | ||
AK129893.1 | - | 5 | 696 | |||
AK291157.1 | - | 5 | 833 | |||
AK291686.1 | - | 5 | 675 | |||
AK300663.1 | - | 6 | 818 | |||
AK304334.1 | - | 4 | 475 | |||
BC039724.1 | - | 5 | 932 | |||
BC106945.1 | - | 5 | 657 | 40034551 | ||
BC106946.1 | - | 5 | 657 | 40034552 | ||
BX647237.1 | - | 4 | 480 | |||
CB162101.1 | - | 3 | 222 | |||
CR749615.1 | - | 5 | 899 |