Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146679.1 | NP_666890.1 | 1 | 520 | ||
GenBank mRNA | BC127962.1 | - | 1 | 524 | 40134881 |