Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146683.1 | NP_666894.1 | 1 | 869 | ||
GenBank mRNA | BC145847.1 | - | 1 | 929 | 40131150 |