Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146708.1 | NP_666919.1 | 1 | 775 | ||
GenBank mRNA | BC116954.1 | - | 1 | 816 | 40126273 |