Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002773.4 | NP_002764.1 | 5 | 857 | ||
GenBank mRNA | AK296656.1 | - | 5 | 576 | ||
AK301619.1 | - | 5 | 660 | |||
BC001462.2 | - | 5 | 768 | 3138532 | ||
L41351.1 | - | 5 | 821 |