Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146725.1 | NP_666936.1 | 1 | 643 | ||
GenBank mRNA | BC114636.1 | - | 1 | 634 |