Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002775.4 | NP_002766.1 | 5, 6 | 1125 | ||
GenBank mRNA | AF070555.1 | - | 3, 4 | 399 | ||
AF097709.1 | - | 5, 6 | 568 | |||
AK091944.1 | - | 7, 8 | 900 | |||
AK092476.1 | - | 6, 7 | 906 | |||
AK290089.1 | - | 5, 6 | 616 | |||
BC011352.1 | - | 6, 7 | 892 | |||
BC031082.1 | - | 5, 6 | 1033 | |||
D87258.1 | - | 5, 6 | 1045 | |||
Y07921.1 | - | 5, 6 | 1035 |